missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LARP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535911
Code : NEW
Additional Details : Weight : 0.00910kg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (91%).
LARP1 Polyclonal antibody specifically detects LARP1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
LARP1 | |
Polyclonal | |
Unconjugated | |
Larp1 | |
0031/08; 0225/29; 1810024J12Rik; 3110040D16Rik; BcDNA:LD02611; CG14065; CG14066; CG42551; CG42551-PA; CG42551-PB; CG42551-PD; CG42551-PE; CG42551-PF; CG42551-PG; CG42551-PH; dlarp; Dmel\CG42551; Dmel_CG42551; drosophila La related protein; KIAA0731; l(3)S003108; l(3)S022529; La related protein; La ribonucleoprotein domain family member 1; La ribonucleoprotein domain family, member 1; La-related protein; la-related protein 1; Larp; Larp1; larp-PA; larp-PB; larp-PD; larp-PE; larp-PF; larp-PG; larp-PH; lethal (3) S003108; lethal (3) S022529; meteor; mKIAA0731; RGD1306683 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23367 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q6PKG0 | |
Larp1 | |
Recombinant fragment corresponding to a region within amino acids 211 and 474 of LARP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |