missing translation for 'onlineSavingsMsg'
Learn More

LAR Mouse, Unlabeled, Clone: 7, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610351

 View more versions of this product

Product Code. 15885409

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

The Leukocyte common Antigen-Related (LAR) receptor protein tyrosine phosphatase (RPTP) is one of approximately 20 known, distinct transmembrane RPTPs. LAR consists of an extracellular region with three Ig-like and eight fibronectin type III-like (FNIII) domains and a cytoplasmic region containing two tyrosine phosphatase domains typical of RPTPs. The Ig and FNIII domains of LAR show some sequence similarity to domains in the neural cell adhesion molecule N-CAM, a neural surface glycoprotein which mediates cell adhesion and neurite outgrowth. Studies suggest that LAR regulates neurite outgrowth and pathfinding during neural development. N-CAM and other cell adhesion molecules that affect neurite outgrowth, such as L1, TAG-1, and fasciculin II, lack intrinsic kinase or phosphatase activity. Thus, LAR is part of a distinct class of proteins with both cell adhesion and tyrosine phosphatase functions. LAR transcripts are alternatively spliced and that splicing is regulated during development. It is postulated that the various spliced forms of LAR each have specialized functions in the nervous system and play a role in neural development and regeneration.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

LAR
Monoclonal
250μg/mL
Leukocyte common Antigen-Related
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
7
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Human, Mouse, Rat
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.