missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-LAMP2a, Polyclonal, Abcam™
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Human, Chinese Hamster and Predicted to work with Dog
Immunogen: Synthetic peptide conjugated to KLH derived from within residues 350 to the C-terminus of Human LAMP2a.
Applications:
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting
Specifications
Specifications
| Antigen | LAMP2a |
| Classification | Polyclonal |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide conjugated to KLH derived from within residues 350 to the C-terminus of Human LAMP2a. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Research Discipline | Cardiovascular, Tags And Cell Markers |
| Primary or Secondary | Primary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?