missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laminin Polyclonal Antibody, DyLight™ 488, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522901
This item is not returnable.
View return policy
Description
This antibody is pan-specific and reacts well with all Laminin isoforms tested: Laminin-1 (alpha-1, beta-1, and gamma-1) and Laminin-2 (alpha-2, beta-1, and gamma-1). The antibody functionally inhibits Laminin in mouse and rat. It binds to Laminin and inhibits most, if not all, of its cell adhesion and growth promotive properties. Immunostaining is enhanced by antigen retrieval with pepsin, especially paraffin tissue. Suggested positive control: basement membranes (blood vessels of any tissue, tumors, nerve).
Laminin antibodies are widely used to label blood vessels and basement membranes.Specifications
Laminin | |
Polyclonal | |
DyLight 488 | |
LAMC1 | |
5830440B04; AA408497; B2e; basement membrane protein; C77966; C80098; C81607; CLM; D130003D08Rik; dy; dystrophia muscularis; LAMA; LAMA1; Lama-1; Lama2; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; Lamb2; LAMC1; Laminin A chain; laminin alpha 2 subunit; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin B2 chain; laminin beta-1 chain; laminin gamma 1 chain; laminin M chain; laminin S; laminin subunit alpha 1; laminin subunit alpha 2; laminin subunit alpha-1; Laminin subunit alpha-2; laminin subunit beta 1; laminin subunit beta-1; laminin subunit gamma 1; laminin subunit gamma-1; laminin, alpha 1; laminin, alpha 2; laminin, beta 1; laminin, beta 2; laminin, gamma 1; laminin, gamma 1 (formerly LAMB2); laminin-1; laminin-1 subunit alpha; Laminin-1 subunit beta; laminin-1 subunit gamma; Laminin-10 subunit beta; laminin-10 subunit gamma; Laminin-11 subunit gamma; laminin-12 subunit alpha; Laminin-12 subunit beta; laminin-2 subunit alpha; Laminin-2 subunit beta; laminin | |
Rabbit | |
IgG fraction | |
RUO | |
100350500, 100357482, 100358679, 100757343, 100761564, 100765555, 101102692, 101105588, 101105743, 101117424, 103346447, 117036, 16772, 16773, 16777, 226519, 284217, 298941, 309368, 316758, 3908, 3912, 3915 | |
4° C, store in dark | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.59 mg/mL | |
50mM sodium borate with 0.05% sodium azide | |
P02468, P02469, P07942, P11047, P19137, P24043, P25391, Q60675 | |
LAMA1, LAMA2, LAMB1, LAMC1 | |
Laminin isolated from mouse EHS tumor. | |
100 μL | |
Primary | |
Hamster, Human, Invertebrate, Mammalia, Mouse, Ovine, Rabbit, Rat | |
Antibody | |
IgG |