missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Thyroxine T4 Mouse anti-Chemical, Clone: T4YCH, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MIT0501
This item is not returnable.
View return policy
Description
MIT0501 targets in ELISA, Western Blot, IP, and RIA applications and shows reactivity with Chemical samples. MIT0501 was formerly sold as a Seradyn product.
Levothyroxine, also L-thyroxine, synthetic T4, or 3,5,3′,5′-tetraiodo-L-thyronine, is a synthetic form of thyroxine (thyroid hormone), used as a hormone replacement for patients with thyroid problems. The natural hormone is chemically in the chiral L-form, as is the pharmaceutical agent. Dextrothyroxine (D-thyroxine) briefly saw research as an anticholesterol agent but was pulled due to cardiac side-effects.
Specifications
| L-Thyroxine T4 | |
| Monoclonal | |
| 1 mg/mL | |
| L-thyroxine; Levothyroxine; Thyroxine; Synthroid; C15H11I4NO4 | |
| Thyroxine-BSA | |
| 1 mg | |
| Primary | |
| Antibody | |
| IgG2b |
| ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
| T4YCH | |
| Unconjugated | |
| Mouse | |
| Protein A | |
| RUO | |
| Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction