Learn More
Ku70 Mouse anti-Hamster, Human, Mouse, Non-human primate, Xenopus laevis, Clone: N3H10, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513110
Description
MA5-13110 targets Ku (p70) in immunofluorescence, immunohistochemistry (paraffin), immunoprecipitation, FACS and Western blot applications and shows reactivity with Human, Non-human primate, and Xenopus laevis samples. Reacts with mouse and hamster in Western blot applications only. The MA5-13110 immunogen is human placental extract designated as PSE1-PL. Antibodies to this protein (and modification) were previously sold as part of a Thermo Scientific Cellomics High Content Screening Kit. This replacement antibody is now recommended for researchers who need an antibody for high content cell based assays. It has been thoroughly tested and validated for cellular immunofluorescence (IF) applications. Further optimization including the selection of the most appropriate fluorescent Dylight conjugated secondary antibody may have to be performed for your high content assay.
The Ku autoantigen is a heterodimer of 70kDa (p70) and ∽80kDa (p80) proteins. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase that phosphorylates certain transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis.
Specifications
| Ku70 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P12956, P23475 | |
| Xrcc6, xrcc6.L | |
| Human placental extract designated as PSE1-PL | |
| 500 μL | |
| Primary | |
| Hamster, Human, Mouse, Non-human Primate, X. laevis | |
| Antibody | |
| IgG2b |
| ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| N3H10 | |
| Unconjugated | |
| XRCC6 | |
| 5′-dRP lyase Ku70, 5′-deoxyribose-5-phosphate lyase Ku70, 70 kDa subunit of Ku antigen, ATP-dependent DNA helicase 2 subunit 1, ATP-dependent DNA helicase II 70 kDa subunit, ATP-dependent DNA helicase II, 70 kDa subunit, CTC box binding factor 75 kDa subunit, CTC box-binding factor 75 kDa subunit, DNA repair protein XRCC6, Ku autoantigen p70 subunit, Ku autoantigen, 70kDa,X-ray repair cross-complementing protein 6, lupus Ku autoantigen protein p70, thyroid autoantigen 70kD (Ku antigen), thyroid | |
| Mouse | |
| Protein A | |
| RUO | |
| 100770214, 14375, 2547, 398357 | |
| 4° C | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.