missing translation for 'onlineSavingsMsg'
Learn More

KRIP-1 Mouse, Unlabeled, Clone: 23, BD

Product Code. 15885579
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15885579 50 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15885579 Supplier BD Biosciences Supplier No. 610680

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Several gene regulatory proteins contain “zinc finger” domains of about 30 amino acids and N-terminal regions of about 75 amino acids, named the Krnppel-associated box A (KRAB-A). The KRAB-A domains have been implicated in transcriptional repression. KRIP-1 (KRAB-A-interacting protein 1) was identified as a result of its interaction with a KRAB-A domain. KRIP-1 is 834 amino acids with an apparent molecular weight of 105kDa. Sequence alignments indicate that KRIP-1 shares identity with TIF-1, a transcriptional regulator of nuclear receptors. The region responsible for the transcriptional activity in KRIP-1 is located in the C-terminal region, which also contains a Cys4-His-Cys3 PHD finger domain.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen KRIP-1
Applications Western Blot
Classification Monoclonal
Clone 23
Concentration 250μg/mL
Conjugate Unconjugated
Description KRAB-A Interacting Protein-1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.