Learn More
KLRG1 Hamster anti-Mouse, NA/LE, Unlabeled, Clone: 2F1, BD
Hamster Monoclonal Antibody
Brand: BD Biosciences 562190
Description
The 2F1 monoclonal antibody specifically binds to KLRG1 (Killer cell Lectin-like Receptor mmunoreceptor Tyrosine-based Inhibitory Motif);its ligand has not beenidentified KLRG1 is expressed mainly as a homodimeric molecule consisting of two N-glycosylated subunits of approximately 30-38 kDa. The level of KLRG1 expression is reduced in MHC class I-deficient mice, although direct binding of KLRG1 to MHC class I antigens could not be detected. Cross-linking of KLRG1 by 2F1 mAb reduces TCR-mediated Ca++mobilization and cytotoxic responses (but not IFN-γ production) by CD8+ T cellsand inhibits IFN-γ and TNF-αproduction and redirected lysis by NK cells.
Specifications
| KLRG1 | |
| Monoclonal | |
| 1.0mg/mL | |
| Klrg1; Killer cell lectin-like receptor subfamily G member 1; MAFA | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2 κ |
| Flow Cytometry | |
| 2F1 | |
| Unconjugated | |
| No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
| A-LAK from C57BL/6 mice | |
| 0.5 mg | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.