missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLK2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human KLK2 protein.
Brand: Abnova H00003817-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MWDLVLSIALSVGCTGAVPLIQSRIVGGWECEKHSQPWQVAVYSHGWAHCGGVLVHPQWVLTAAHCLKKNSQVWLGRHNLFEPEDTGQRVPVSHSFPHPLYNMSLLKHQSLRPDEDSSHDLMLLRLSEPAKITDVVKVLGLPTQEPALGTTCYASGWGSIEPEEFLRPRSLQCVSLHLLSNDMCARAYSEKVTEFMLCAGLWTGGKDTCGVSHPYSQHLEGKGSpecifications
| KLK2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human KLK2 protein. | |
| KLK2 | |
| KLK2A2/MGC12201/hK2 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 3817 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| No additive | |
| NM_001002231.1 | |
| KLK2 | |
| KLK2 (NP_001002231.1, 1 a.a. ∼ 223 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction