missing translation for 'onlineSavingsMsg'
Learn More

anti-KLH, APC, Clone: B81-3, BD

Product Code. 15861699
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15861699 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15861699 Supplier BD Biosciences Supplier No. 562169
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Host Species: Hamster
Clone: B81-3
Isotype: IgG2, κ
Immunogen: Keyhole Limpet Hemocyanin

Flow Cytometry, Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Antigen KLH
Classification Monoclonal
Clone B81-3
Concentration 0.2mg/mL
Conjugate APC
Host Species Hamster
Immunogen Keyhole Limpet Hemocyanin
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.