missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kruppel-like factor 3 (basic), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant KLF3.
Brand: Abnova H00051274-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: MLMFDPVPVKQEAMDPVSVSYPSNYMESMKPNKYGVIYSTPLPEKFFQTPEGLSHGIQMEPVDLTVNKRSpecifications
| Kruppel-like factor 3 (basic) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant KLF3. | |
| KLF3 | |
| BKLF/MGC48279 | |
| Mouse | |
| 50 μL | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_016531 | |
| KLF3 | |
| KLF3 (NP_057615, 1 a.a. ∼ 69 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 51274 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction