Learn More
Kruppel-like factor 13, Mouse, Clone: 1D3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant KLF13.
Brand: Abnova H00051621-M01.100ug
Description
KLF13 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes (Scohy et al., 2000 [PubMed 11087666]).[supplied by OMIM
Sequence: PESRPEGAAVAATPTLPRVEERRDGKDSASLFVVARILADLNSpecifications
| Kruppel-like factor 13 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC010438 | |
| KLF13 | |
| KLF13 (AAH13946, 31 a.a. ∼ 72 a.a) partial recombinant protein. | |
| 100 μg | |
| Yes | |
| 51621 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Western Blot | |
| 1D3 | |
| Mouse monoclonal antibody raised against a partial recombinant KLF13. | |
| KLF13 | |
| BTEB3/FKLF2/NSLP1/RFLAT-1/RFLAT1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.