missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR3DL1 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
£167.00 - £541.00
Specifications
| Antigen | KIR3DL1 |
|---|---|
| Clone | DX9 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18765238
![]() |
R&D Systems
FAB1225A-025 |
25 Tests |
£167.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18746996
![]() |
R&D Systems
FAB1225A-100 |
100 Tests |
£541.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIR3DL1 Monoclonal specifically detects KIR3DL1 in Human samples. It is validated for Flow Cytometry.Specifications
| KIR3DL1 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 3811 | |
| Human KIR3DL1-expressing NK cell clone | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| DX9 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human | |
| AMB11, CD158 antigen-like family member E, CD158E, CD158e antigen, CD158e1, CD158e1/2, CD158e2, cl-11, cl-2, HLA-BW4-specific inhibitory NK cell receptor, killer cell immunoglobulin-like receptor 3DL1, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail1,nkat3, killer Ig receptor, KIR antigen 3DL1, KIR3DL1/S1, KIR3DS1, MGC119726, MGC119728, MGC126589, MGC126591, MHC class I NK cell receptor, Natural killer-associated transcript 3, Nkat3, NKAT-3, NKAT3KIR, NKB1, NKB1B, NKB1CD158E1, NK-receptor, p70 killer cell inhibitory receptor, p70 natural killer cell receptor clones CL-2/CL-11, p70 NK receptor CL-2/CL-11 | |
| KIR3DL1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human KIR3DL1. Did not cross-react with other KIR family members when tested against a series of KIR-expressing BaF/3 tansfectants (KIR2DL1, KIR2DL2, KIR2DL3, KIR2DL4, KIR2DS1, KIR2DS2, KIR2DS3, KIR2DS4, and KIR3DL2; Bakker, A.B.H. et al. (1998) J. Immunol. 160:5239). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title