missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human KIR2DL4/CD158d PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
£190.00 - £421.00
Specifications
| Antigen | KIR2DL4/CD158d |
|---|---|
| Clone | 181703 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18799328
![]() |
R&D Systems
FAB2238P-025 |
25 Tests |
£190.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18768206
![]() |
R&D Systems
FAB2238P-100 |
100 Tests |
£421.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIR2DL4/CD158d Monoclonal specifically detects KIR2DL4/CD158d in Human samples. It is validated for Flow Cytometry.Specifications
| KIR2DL4/CD158d | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 103AS, 15.212, CD158 antigen-like family member D, CD158d, CD158d antigen, CD158DKIR103ASKiller cell inhibitory receptor 103AS, G9P, killer cell immunoglobulin-like receptor 2DL4, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 4, killer Ig receptor, KIR103, KIR-103AS, MHC class I NK cell receptor KIR103AS, natural killer cell inhibitory receptor, NK cell receptor | |
| KIR2DL4 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Stains human KIR2DL4 transfected BaF/3 cells. It does not recognize transfectants that express KIR2DL1, 2DL2, 2DL3, 2DL5, 2DS1, 2DS2, 2DS4, 3DL1, 3DL2, or 3DS1. |
| 181703 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| Q99706 | |
| 3805 | |
| BaF3 mouse pro-B cell line transfected with human KIR2DL4 Accession # Q99706 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title