Learn More
KIR-NKAT2 Mouse anti-Human, FITC, Clone: DX27, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 556070
Description
Reacts with KIR-NKAT2, a member of the killer-inhibitory receptor (KIR) family. The KIR family consists of transmembrane glycoproteins of the Ig superfamily expressed on NK cells and a subset of T cells which are involved in recognition of MHC class I molecules on target cells and inhibit cytotoxicity. KIR-NKAT2 antigen is involved in the recognition of HLA-C molecules (-Cw3 and related alleles).
Host Species: Mouse
Clone: DX27
Isotype: IgG2a κ
Species Reactivity: Human
Flow Cytometry
Specifications
| KIR-NKAT2 | |
| Monoclonal | |
| FITC | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| DX27 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.