Learn More
ketohexokinase (fructokinase), Mouse, Clone: 1H6-2B6, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant KHK.
Brand: Abnova H00003795-M01.100ug
Description
This gene encodes ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The product of this gene is the first enzyme with a specialized pathway that catabolizes dietary fructose. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Sequence: MEEKQILCVGLVVLDVISLVDKYPKEDSEIRCLSQRWQRGGNASNSCTILSLLGAPCAFMGSMAPGHVADFVLDDLRRYSVDLRYTVFQTTGSVPIATVIINEASGSRTILYYDRSLPDVSATDFEKVDLTQFKWIHIEGRNASEQVKMLQRIDAHNTRQPPEQKIRVSVEVEKPREELFQLFGYGDVVFVSKDVAKHLGFQSAEEALRGLYGRVRKGAVLVCAWAEEGADALGPDGKLLHSDAFPPPRVVDTLGAGDTFNASVIFSLSQGRSVQEALRFGCQVAGKKCGLQGFDGIVSpecifications
| ketohexokinase (fructokinase) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC006233 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 3795 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Immunohistochemistry (PFA fixed), KnockDown, Western Blot | |
| 1H6-2B6 | |
| Mouse monoclonal antibody raised against a full length recombinant KHK. | |
| KHK | |
| KHK | |
| KHK (AAH06233, 1 a.a. ∼ 298 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.