Learn More
Invitrogen™ KEAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534454
Description
A suggested positive control is human lung tissue lysate. PA5-34454 can be used with blocking peptide PEP-1496. The PA5-34454 immunogen is located within the last 50 amino acids of KEAP1. Predicted molecular ~ 70kD. In Western blot applications, this antibody has been observed to detect a band at: 65-70kD. Predicted species reactivity based on immunogen sequence: Pig: (100%).
This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.
Specifications
| KEAP1 | |
| Polyclonal | |
| Unconjugated | |
| Keap1 | |
| cytosolic inhibitor of Nrf2; INrf2; Keap1; kelch like ECH associated protein 1; Kelch-like ECH-associated protein 1; kelch-like family member 19; kelch-like protein 19; Kiaa0132; KLHL19; mKIAA0132; NRF2 cytosolic inhibitor; ring canal protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117519, 50868, 9817 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P57790, Q14145, Q9Z2X8 | |
| Keap1 | |
| A peptide near the carboxy terminus of human KEAP1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.