missing translation for 'onlineSavingsMsg'
Learn More

Karyopherin α Mouse, Unlabeled, Clone: 2, BD

Mouse Monoclonal Antibody

£151.41 - £428.00

Specifications

Antigen Karyopherin α
Clone 2
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15815489
View Documents
BD Biosciences
610485
50 μg
ONLINE EXCLUSIVE PRICE
£158.00
£151.41 / 1 set
Save £6.59
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15825489
View Documents
BD Biosciences
610486
150 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

The two step process of importing proteins into the nucleus involves the binding and interaction of several cytosolic and nuclear pore proteins. Proteins to be translocated into the nucleus contain a nuclear localization sequence (NLS) which is recognized and bound by carrier proteins in the cytosol. Heterodimers belonging to a highly conserved family of proteins called karyopherins are required for successful nuclear localization of cytosolic proteins. The α-subunits appear to function in the binding of NLS (both simple and bitartite NLS motifs), but both α- and β-subunits are required for successful docking to the nuclear envelope. ATP is required for complete translocation of proteins into the nucleus. Karyopherin α2 was first identified as Rch-1, an NLS receptor which interacts with the RAG-1 recombination-activating protein in developing B and T cells. Rch-1 has been reported to be 44% identical to karyopherin α1 (hSRP-1 /NPI-1).

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Karyopherin α
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
Rch-1
2
Western Blot
Unconjugated
RUO
Canine, Drosophila, Human, Mouse, Rat
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.