Learn More
Invitrogen™ Mouse anti-Human Kappa Light Chain Secondary Antibody
Mouse Monoclonal Secondary Antibody
Brand:  Invitrogen™ MA110385
Product Code. 12342393
  • £245.00 / Pack of 100
Description
This antibody reacts with kappa light chains (22.5 kDa) of immunoglobulins. This antibody will not cross-react with hamster, goat, guinea pig, rabbit or sheep.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human Kappa Light Chain
Monoclonal
1 mg/mL
PBS with 15mM sodium azide; pH 7.4
not available.
100 μg
Secondary
4°C, do not freeze
Liquid
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
A8B5
Unconjugated
Mouse
Protein A
RUO
Human
Secondary Antibody
IgG1