Learn More
K+ Channel α Mouse, Unlabeled, Clone: 32, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 611249
Description
Cellular excitability is modulated by the precise function of voltage-sensitive ion channels. Large conductance potassium channels (Maxi-K) are unique in that they are sensitive to transmembrane potential and intracellular Ca2+; concentrations. These channels, important for neuronal firing and vascular tone, share many features with voltage-dependent Na+, Ca2+;, and K+ channels. Among these are the S4 region, a motif with a repeated triple sequence of one positively charged amino acid and two hydrophobic amino acids. This region is thought to be the voltage sensor. Maxi-K is subject to complex metabolic control control that also involves G proteins and phosphorylation/dephosphorylation reactions. This type of K+ channel is composed of two subunits, the pore-forming α subunit (hslo) and the regulatory β subunit. The α subunit is subject to direct phosphorylation by cyclic GMP-dependent protein kinase (PKG) and dephosphorylation by protein phosphatase 2A. The fact that the α subunit of Maxi-K channels is a substrate of PKG supports the idea that these channels perform an important function in the cellular response to potent vasodilators, such as nitrocompounds and atrial natiuretic peptide.
Host Species: Mouse
Clone: 32
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Human K+ Channel α Subunit aa. 995-1113
Formula Weight [Chemical]: 125kDa
Immunofluorescence, Western Blotting
Specifications
| K+ Channel α | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Human K+ Channel α Subunit aa. 995-1113 | |
| 150 μg | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 32 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.