missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ JMJD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544862
This item is not returnable.
View return policy
Description
Immunogen is a synthetic peptide within LKQDISIPDY CSLGDGEEEE ITINAWFGPQ GTISPLHQDP QQNFLVQVMG. Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
This gene likely encodes a histone lysine demethylase. Studies of a similar protein in mouse indicate a potential role for this protein as a tumor suppressor. Alternatively spliced transcript variants have been described.
Specifications
| JMJD5 | |
| Polyclonal | |
| Unconjugated | |
| Kdm8 | |
| 3110005O21Rik; arginyl C3-hydroxylase KDM8; Bifunctional peptidase and arginyl-hydroxylase JMJD5; bifunctional peptidase and arginyl-hydroxylase JMJD5; jmjC domain-containing protein 5; hypothetical protein FLJ13798; jmjC domain-containing protein 5; Jmjd5; Jumonji C domain-containing protein 5; jumonji domain containing 5; jumonji domain-containing protein 5; Kdm8; L-arginine (3R)-hydroxylase KDM8; LOW QUALITY PROTEIN: lysine-specific demethylase 8; lysine (K)-specific demethylase 8; lysine demethylase 8; lysine-specific demethylase 8; RGD1304823; zgc:173863; zgc:91962 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79831 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N371 | |
| Kdm8 | |
| Synthetic peptide directed towards the middle region of human JMJD5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction