Learn More
junctional adhesion molecule 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human JAM2 protein.
Brand: Abnova H00058494-D01P.100ug
Description
Tight junctions represent one mode of cell-to-cell adhesion in epithelial or endothelial cell sheets, forming continuous seals around cells and serving as a physical barrier to prevent solutes and water from passing freely through the paracellular space. The protein encoded by this immunoglobulin superfamily gene member is localized in the tight junctions between high endothelial cells. It acts as an adhesive ligand for interacting with a variety of immune cell types and may play a role in lymphocyte homing to secondary lymphoid organs. [provided by RefSeq
Sequence: MARRSRHRLLLLLLRYLVVALGYHKAYGFSAPKDQQVVTAVEYQEAILACKTPKKTVSSRLEWKKLGRSVSFVYYQQTLQGDFKNRAEMIDFNIRIKNVTRSDAGKYRCEVSAPSEQGQNLEEDTVTLEVLVAPAVPSCEVPSSALSGTVVELRCQDKEGNPAPEYTWFKDGIRLLENPRLGSQSTNSSYTMNTKTGTLQFNTVSKLDTGEYSCEARNSVGYRRCPGKRMQVDDLNISGIIAAVVVVALVISVCGLGVCYAQRKGYFSKETSFQKSNSSSKATTMSENDFKHTKSFIISpecifications
| junctional adhesion molecule 2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human JAM2 protein. | |
| JAM2 | |
| C21orf43/CD322/JAM-B/JAMB/PRO245/VE-JAM/VEJAM | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_021219.2 | |
| JAM2 | |
| JAM2 (NP_067042.1, 1 a.a. ∼ 298 a.a) full-length human protein. | |
| 100 μg | |
| Cell Adhesion | |
| 58494 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.