missing translation for 'onlineSavingsMsg'
Learn More

Anti-JAK2 (phospho Y1007 + Y1008) + JAK1 (phospho Y1034 + Y1035) antibody [E132], Abcam

Product Code. 32076499 Shop All Abcam Products
Change view
Click to view available options
Quantity:
20 μL
Unit Size:
20µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32076499 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32076499 Supplier Abcam Supplier No. ab3210120uL
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Rabbit Recombinant Monoclonal JAK2 phospho Y1007 + Y1008 antibody. Suitable for IHC-P, ELISA, Dot, WB, ICC/IF, Flow Cyt (Intra) and reacts with Human, Synthetic peptide, Mouse, Rat samples. Cited in 304 publications.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen JAK2 pY1007 + Y1008 (JAK2)
Applications Dot Blot, ELISA, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry (FFPE), Western Blot
Cellular Localization Endomembrane system; Peripheral membrane protein; Cytoplasm; Nucleus
Classification Recombinant Monoclonal
Clone E132
Conjugate Unconjugated
Formulation pH: 7.2 - 7.4, Preservative: 0.01% Sodium azide, Constituents: PBS, 50% Glycerol (glycerin, glycerine), 0.05% BSA
Gene JAK2
Gene Accession No. O60674
Host Species Rabbit
Immunogen JAK2 pY1007 + Y1008 (JAK2)
Purification Method Affinity purification
Quantity 20 μL
Regulatory Status RUO
Research Discipline Immunology & Infectious Disease
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG
Shipping Condition Conditional Ambient
Target Modification State Phospho-Y1007∣Phospho-Y1008
Target Gene JAK2
Target Molecule JAK2 pY1007 + Y1008 (JAK2)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.