Learn More
IRS-1 (pY896) Mouse, PE, Clone: K9-211, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558440
Description
The IRS (Insulin Receptor Substrate) proteins IRS-1, IRS-2, IRS-3, and IRS-4 are major substrates of the insulin receptor and the insulin-like growth factor-1 (IGF-1) receptor tyrosine kinases. IRS proteins contain an N-terminal pleckstrin homology (PH) domain, a phosphotyrosine-binding (PTB) domain, and multiple tyrosine phosphorylation sites in the C-terminus. The IRS-1 protein is widely expressed and, along with IRS-2, mediates somatic growth and carbohydrate metabolic responses to insulin. Following insulin receptor ligation, IRS-1 binds to the juxtamembrane region of the receptor via the PH and PTB domains and is tyrosine phosphorylated, which facilitates its interaction with SH2 domain-containing signaling proteins. Specifically, the phosphorylated tyrosine 896 (pY896) of human IRS-1 is a major binding site for the GRB2 (Growth-factor Receptor-Bound protein 2) adaptor protein. After IRS-1 activation, negative and positive feedback regulates dephosphorylation of its tyrosine sites, which ultimately regulates the magnitude and/or duration of the downstream pleiotropic responses to insulin and IGF-1. The K9-211 monoclonal antibody recognizes pY896 of human IRS-1. The orthologous phosphorylation sites of mouse and rat IRS-1 are Y891 and Y895, respectively.
Intracellular Staining
Specifications
| IRS-1 (pY896) | |
| Monoclonal | |
| PE | |
| Mouse | |
| 50 Tests | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG2a κ |
| Flow Cytometry | |
| K9-211 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Phosphorylated Human IRS-1 Peptide | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.