missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IRS1 Monoclonal Antibody (J.566.7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515072
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
IRS-1, a major substrate of the insulin receptor, is phosphorylated in response to stimulation of cells by insulin, insulin-like growth factor 1 (IGF-1) and interleukin 4 (IL-4). IRS-1 is phosphorylated on serine, threonine and tyrosine residues in a variety of tissues. An insulin-sensitive serine/threonine kinase casein kinase II mediates a portion of the insulin-stimulated serine/threonine phosphorylation of overexpressed IRS-1 in vivo. Thr 502 is identified as the major casein kinase II-catalyzed phosphorylation site in rat IRS-1, and Ser 99 is an additional phosphorylation site catalyzed by casein kinase II. Thus, casein kinase II-catalyzed phosphorylation of IRS-1 may be a component of the intracellular insulin signaling cascade. IRS-1 contains three putative binding sites for 14-3-3 (Ser 270, Ser 374 and Ser 641) and the motif around Ser 270 is located in the phosphortyrosine binding domain of IRS-1, which is responsible for the interaction with the insulin receptor.
Specifications
| IRS1 | |
| Monoclonal | |
| 708 μg/mL | |
| 0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| P35568, P35569, P35570 | |
| IRS1 | |
| Synthetic peptide corresponding to the sequence surrounding Ser612 of human IRS-1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG1 |
| Western Blot | |
| J.566.7 | |
| Unconjugated | |
| IRS1 | |
| G972R; HIRS-1; Insulin receptor substrate 1; IRS1; IRS-1; IRS1IRM; pp185 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 16367, 25467, 3667 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction