missing translation for 'onlineSavingsMsg'
Learn More

IRS-1 Mouse, Unlabeled, Clone: 6, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611394

 View more versions of this product

Product Code. 15815919

  • ONLINE EXCLUSIVE PRICE
    £193.00
    £185.50 / Each
    Save £7.50
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The IRS (Insulin receptor substrate) proteins IRS-1, IRS-2, IRS-3, and IRS-4 are major substrates of the insulin receptor tyrosine kinase and the insulin-like growth factor-1 receptor. IRS proteins contain an N-terminal pleckstrin homology (PH) domain, an ATP-binding domain, and multiple tyrosine phosphorylation sites in the C-terminus. Following insulin receptor ligation, IRS-1 binds to the juxtamembrane region of the receptor and is tyrosine phosphorylated. This facilitates its interaction with SH2 domain-containing signaling proteins, such as PI3 kinase, fyn, Grb2, and PTP1D. Phosphorylation dramatically reduces the affinity of IRS-1 for the insulin receptor, indicating that dissociation from the receptor and subsequent subcellular translocation are important to IRS-1 function in the pleiotropic effects induced by insulin. In support of this, IRS-1-null mice are viable, but exhibit growth retardation and abnormal glucose metabolism. In cases of reduced IRS-1 expression, certain IRS-1 functions can be assumed by the related IRS-2 protein, while other activities linked to IRS-1 are inhibited. Thus, IRS-1 is an essential component of insulin induced signal transduction.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IRS-1
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Rat IRS-1 aa. 1131-1234
50 μg
Cell Biology
Mouse, Rat
IgG1
Western Blot
6
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.