Learn More
interferon regulatory factor 3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human IRF3 protein.
Brand: Abnova H00003661-B01.50uL
Description
IRF3 encodes interferon regulatory factor 3, a member of the interferon regulatory transcription factor (IRF) family. IRF3 is found in an inactive cytoplasmic form that upon serine/threonine phosphorylation forms a complex with CREBBP. This complex translocates to the nucleus and activates the transcription of interferons alpha and beta, as well as other interferon-induced genes. [provided by RefSeq
Sequence: MGTPKPRILPWLVSQLDLGQLEGVAWVNKSRTRFRIPWKHGLRQDAQQEDFGIFQAWAEATGAYVPGRDKPDLPTWKRNFRSALNRKEGLRLAEDRSKDPHDPHKIYEFVNSGVGDFSQPDTSPDTNGGGSTSDTQEDILDELLGNMVLAPLPDPGPPSLAVAPEPCPQPLRSPSLDNPTPFPNLGPSENPLKRLLVPGEEWEFEVTAFYRGRQVFQQTISCPEGLRLVGSEVGDRTLPGWPVTLPDPGMSLTDRGVMSYVRHVLSCLGGGLALWRAGQWLWAQRLGHCHTYWAVSEELLPNSGHGPDGEVPKDKEGGVFDLGPFIVDLITFTEGSGRSPRYALWFCVGESWPQDQPWTKRLVMVKVVPTCLRALVEMARVGGASSLENTVDLHISNSHPLSLTSDQYKAYLQDLVEGMDFQGPGESSpecifications
| interferon regulatory factor 3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human IRF3 protein. | |
| IRF3 | |
| IRF3 | |
| IRF3 (NP_001562.1, 1 a.a. ∼ 427 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 3661 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| No additive | |
| NM_001571.2 | |
| Mouse | |
| 50 μL | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.