missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Mouse anti-Human, Clone: 17C2, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 392700
This item is not returnable.
View return policy
Description
Interferons (IFN)s are involved in a multitude of immune interactions during viral infections and play a major role in both the induction and regulation of innate and adaptive antiviral mechanisms. During infection, host-virus interactions signal downstream molecules such as transcription factors such as IFN regulatory factor-3 (IRF3) which can act to stimulate transcription of IFN-alpha/beta genes. IRF3 is present in an inactive form in the cytoplasm of most cells. Following viral infection, IRF3 can be activated by Ikappa-B kinase-iota and TANK-binding kinase 1 (TBK1), whereupon IRF3 translocates to the nucleus. IRF3 can also be activated by stimulation of toll-like receptor 3 (TLR3) by dsRNA. IRF3 exists as at least two distinct isoforms.
Specifications
| IRF3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| IRF3 | |
| IRF3, Interferon regulatory factor 3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3661 | |
| -20°C | |
| Liquid |
| ELISA, Immunofluorescence, Western Blot | |
| 17C2 | |
| Unconjugated | |
| Q14653 | |
| IRF3 | |
| Recombit protein derived from the C-terminal region of the human IRF3 (interferon regulatory factor 3) protein | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction