Learn More
interleukin-1 receptor-associated kinase 2, Mouse, Clone: 3H1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant IRAK2.
Brand: Abnova H00003656-M03.100ug
Description
IRAK2 encodes the interleukin-1 receptor-associated kinase 2, one of two putative serine/threonine kinases that become associated with the interleukin-1 receptor (IL1R) upon stimulation. IRAK2 is reported to participate in the IL1-induced upregulation of NF-kappaB. [provided by RefSeq
Sequence: KPEKPLAASVRKAEDEQEEGQPVRMATFPGPGSSPARAHQPAFLQPPEEDAPHSLRSDLPTSSDSKDFSTSIPKQEKLLSLAGDSLFWSEADVVQATDDFSpecifications
| interleukin-1 receptor-associated kinase 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001570 | |
| IRAK2 | |
| IRAK2 (NP_001561, 111 a.a. ∼ 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3H1 | |
| Mouse monoclonal antibody raised against a partial recombinant IRAK2. | |
| IRAK2 | |
| IRAK-2/MGC150550 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 3656 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.