missing translation for 'onlineSavingsMsg'
Learn More

IQGAP1 Mouse, Unlabeled, Clone: 24, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610612

 View more versions of this product

Product Code. 15895539

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

GTPases (Ras, Rho, cdc42Hs, and rac) modulate various cellular functions such as cytoskeletal architecture, growth, motility, and gene expression. The activity of the GTP-binding proteins is regulated by factors that accelerate GTP-binding (GAPs) and proteins that enhance the rate of GTP hydrolysis (GTPases). IQGAP1 is Ras-GAP related protein that contains a WW domain that binds poly-proline regions, a calponin-homology domain (CHD), an IQ domain of unconventional myosins, and a C-terminal GAP-related domain (GRD). IQGAP1 co-immunoprecipitates with cdc42 and calmodulin, and colocalizes with cdc42Hs at the leading edges of the plasma membrane and at the cell-cell junctions. IQGAP1 may interact with both cadherins and catenins leading to dissociation of these proteins from sites of cell adhesion. Thus, IQGAP1 may be a Rac1 and Cdc42 effector that negatively regulates cadherin-mediated cell junctions.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IQGAP1
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG1
Western Blot
24
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.