Learn More
CXCL10 Monoclonal Antibody (4D5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180897
Code : LD
Additional Details : Weight : 0.01000kg
Description
Mouse anti Human IP-10 antibody, clone 4D5 recognizes human C-X-C motif chemokine 10, also known as IP-10, Small-inducible cytokine B10 or CXCL10. IP-10 is a 98 amino acid approximately 10kDa CxC chemokine induced by interferon gamma (P02778).
CXCL10 Monoclonal antibody specifically detects CXCL10 in Human samples. It is validated for ELISA, Western Blot.Specifications
CXCL10 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P02778 | |
CXCL10 | |
Recombinant human IP-10 | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Western Blot | |
4D5 | |
Unconjugated | |
CXCL10 | |
10 kDa interferon gamma-induced protein; 10 kDa interferon-gamma induced protein precursor; C Cmotif chemokine; C X C motif chemokine; C7; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 10; chemokine C-X-C motif ligand 10; chemokine CXC-like protein; Crg2; CRG-2; CXC; CXC chemokine; CXC motif chemokine; C-X-C motif chemokine 10; C-X-C motif chemokine 10-like protein; C-X-C motif chemokine ligand 10; CXCL; CXCL10; CXCL10(1-73); gamma interferon inducible protein 10; gamma IP10; gamma-IP10; gIP-10; H-IP-10; IFI10; Inp10; interferon activated gene 10; interferon-gamma induced protein CRG-2; interferon-gamma-induced protein CRG-2; interferon-gamma-inducible protein 10; interferon-gamma-inducible protein-10; interferon-inducible cytokine IP-10; interferon-inducible protein 10; IP10; IP-10; M-IP-10; Mob1; mob-1; protein 10 from interferon (gamma)-induced cell line; protein Mob-1; Scyb10; similar to Small inducible cytokine B10 precursor (CXCL10) (10 kDa interferon-gamm | |
Mouse | |
Protein A | |
RUO | |
3627 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |