Learn More
CXCL10 Rabbit anti-Mouse, Biotin, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA132923
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-32923 detects IP10 from mouse samples. PA1-32923 has been successfully used in Western blot applications. The PA1-32923 immunogen is: Recombit mouse IP-10 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.Specifications
CXCL10 | |
Polyclonal | |
Cxcl10 | |
C7, CRG-2, INP10, IP-10, IP10, Ifi10, Scyb10, gIP-10, mob-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
15945 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
Biotin | |
P17515 | |
CXCL10 | |
Recombit mouse IP-10 | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |