Learn More
IP-10 Mouse anti-Human, Unlabeled, Clone: 4D5, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555046
Description
The monoclonal antibody 4D5/A7/C5 reacts with human CXC chemokine, interferon gamma inducible protein (IP-10). IP-10 is inducible in monocytes, keratinocytes, and endothelial cells by IFN-γ. This antibody is routinely tested by ELISA Capture. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
ELISA
Specifications
| IP-10 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human IP-10 protein | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG2a κ |
| ELISA | |
| 4D5 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.