Learn More
Invariant NK T Cell Mouse anti-Human, FITC, Clone: 6B11, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558371
Description
Monoclonal antibody 6B11 reacts with a unique determinant in the CDR3 region of the invariant (Vα24-JαQ) TCR chain. Thus 6B11 identifies a subset of all Vα24 positive T cells. The binding of the PE-conjugate of 6B11 can be partially blocked by other antibodies specific for Vα24 (BD Biosciences Pharmingen, data not shown). This invariant chain is expressed on invariant NK T cells, which are a small subset of T lymphocytes that also express other NK cell molecules such as CD161. Invariant NK T cells have been reported to play an immunoregulatory role. The Vα24-JαQ interacts with the CD1d protein. Invariant NK T cells can be specifically activated by CD1d-presented antigen resulting in rapid production of IL-4 and IFN (also known as IFN-γ). Studies of NK T cells describe their role in anti-tumor immunity and autoimmunity. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry
Specifications
| Invariant NK T Cell | |
| Monoclonal | |
| FITC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 6B11 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Peptide from CDR3 loop of the invariant TCR V24 sequence | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.