missing translation for 'onlineSavingsMsg'
Learn More

CD51 Mouse anti-Human, Clone: 272-17E6, Invitrogen™

Product Code. 11515622
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11515622 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11515622 Supplier Invitrogen Supplier No. MA191669

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-91669 detects Integrin alpha V from human samples. MA1-91669 has been successfully used in ELISA, flow cytometry, immunohistochemistry, immunoprecipitation applications. The MA1-91669 immunogen is: Full length native protein (purified) (Human). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD51
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 272-17E6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Gene ITGAV
Gene Accession No. P06756
Gene Alias CD51, MSK8, VNRA, antigen identified by monoclonal antibody L230, integrin alpha-V, vitronectin receptor subunit alpha, ITGAV
Gene Symbols ITGAV
Host Species Mouse
Immunogen Full length native protein (purified) (Human).
Purification Method IgG fraction
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3685
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.