missing translation for 'onlineSavingsMsg'
Learn More

anti-Integrin alpha V/CD51 PE, Clone: RMV-7, Invitrogen™

Integrin alpha V/CD51 Monoclonal antibody A16332 from Invitrogen specifically detects Integrin alpha V/CD51 in Mouse samples. It is validated for Flow Cytometry.

Brand:  Invitrogen A16332

Product Code. 15296014

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Integrin alpha V/CD51 Monoclonal antibody A16332 from Invitrogen specifically detects Integrin alpha V/CD51 in Mouse samples. It is validated for Flow Cytometry.

ITAGV encodes integrin alpha chain V. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. The I-domain containing integrin alpha V undergoes post-translational cleavage to yield disulfide-linked heavy and light chains, that combine with multiple integrin beta chains to form different integrins. Among the known associating beta chains (beta chains 1,3,5,6, and 8; 'ITGB1', 'ITGB3', 'ITGB5', 'ITGB6', and 'ITGB8'), each can interact with extracellular matrix ligands; the alpha V beta 3 integrin, perhaps the most studied of these, is referred to as the Vitronectin receptor (VNR). In addition to adhesion, many integrins are known to facilitate signal transduction. Alternative splicing results in multiple transcript variants.
Host Species: Rat
Cross Reactivity: Mouse
Isotype: IgG1, κ
Clone: RMV-7
Immunogen: mouse CD51
Formulation: 50μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: CD51, MSK8, VNRA, antigen identified by monoclonal antibody L230, integrin alpha-V, vitronectin receptor subunit alpha, ITGAV
Uniprot Accession: P43406
Entrez GeneID: 16410
Gene Symbol: Itgav
This product detects Integrin alpha V/CD51 in Mouse samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is mouse CD51.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Integrin alpha V/CD51
Monoclonal
PE
P43406
Itgav
mouse CD51
Primary
Mouse
Purified
Flow Cytometry
RMV-7
50μg Purified Antibody (0.2mg/ml) in proprietary buffer, with 0.09% sodium azide
CD51, MSK8, VNRA, antigen identified by monoclonal antibody L230, integrin alpha-V, vitronectin receptor subunit alpha, ITGAV
Rat
50 μg
16410
Store at 4° C
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.