Learn More
anti-Insulin Receptor, Clone: MA-20, Invitrogen™
Specifically detect CD220 / Insulin Receptor in Human samples using Thermo Scientifc™ Pierce™ anti-CD220 / Insulin Receptor Clone: MA-20, validated for BLOCK, Neu and WB
Brand: Thermo Scientific MA1-16812
Description
Specifically detect CD220 / Insulin Receptor in Human samples using Thermo Scientifc™ Pierce™ anti-CD220 / Insulin Receptor Clone: MA-20, validated for BLOCK, Neu and WB
The insulin receptor is functionally important for negative cooperativity, down-regulation, and beta-subunit tyrosine kinase activity. There are two recognized epitopes of the alpha-subunit. One epitope is close to the insulin-binding site and plays a role in insulin-induced negative cooperativity and down-regulation. The other site (recognized by the monoclonal MA-20 antibody) is at a distance from the insulin-binding site, where it only plays a role in down-regulation. Both epitopes appear to be independent of insulin receptor tyrosine kinase activity.
Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG2b
Clone: MA-20
Immunogen: Highly purified insulin receptors (alpha-subunit) from human term placentas.
Formulation: 100μL of Protein G purified antibody (1.2mg/mL) in PBS buffer; pH 7.4.
Uniprot Accession: P06213
Gene Symbol: INSR
Entrez Gene ID: 3643, 24954
MA1-16812 detects CD220 / Insulin Receptor in Human samples. MA1-16812 has been successfully used in BLOCK, Neu and WB procedures.MA1-16812 The immunogen is Highly purified insulin receptors (alpha-subunit) from human term placentas.
Specifications
| Insulin Receptor | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| 100 μL | |
| Primary | |
| Store at -20°C. Avoid freeze/Thaw cycles. | |
| IgG2b |
| ELISA, Immunoprecipitation | |
| MA-20 | |
| INSR | |
| Highly purified insulin receptors (alpha-subunit) from human term placentas. | |
| RUO | |
| Human | |
| Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.