missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ING5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521443
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ING5 is similar to ING1, a tumor suppressor protein that can interact with TP53, inhibit cell growth, and induce apoptosis. This protein contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This protein can bind TP53 and EP300/p300, a component of the histone acetyl transferase complex, suggesting its involvement in TP53-dependent regulatory pathway.
Specifications
| ING5 | |
| Polyclonal | |
| Unconjugated | |
| ING5 | |
| 1700001C14Rik; 1700027H23Rik; 1810018M11Rik; AI225768; ING5; inhibitor of growth family member 5; inhibitor of growth family, member 5; inhibitor of growth protein 5; p28ING5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84289 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
| Q8WYH8 | |
| ING5 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human ING5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction