Learn More
Invitrogen™ Influenza A H1N1 Virions Polyclonal Antibody, HRP

Goat Polyclonal Antibody
Brand: Invitrogen™ PA173045
Description
The PA1-73045 antibody reacts with Influenza A virions H1N1. It does not react with Influenza B, RSV, Para 1-3 or adeno viruses. PA1-73045 has been successfully used in ELISA applications. The PA1-73045 antibody was raised against Influenza A, strain USSR (H1N1). NOTE: sodium azide is a potent inhibitor of peroxidase.
Influenza viruses consists of three types (Influenza A, B, and C), which make up three of the five genera in the Orthomyxoviridae family. Commonly know as the flu, influenza strains can affect both birds and mammals. Influenza strains form roughly spherical particles and are similar in composition with viral envelope containing two main types of glycoproteins (hemagglutinin and neuraminidase), wrapped around the central core of the viral RNA genome and other viral proteins that package and protect the RNA. Unusually for a virus, the influenzavirus genome is not a single piece of nucleic acid; rather a seven to eight segmented fragments of negative-sense RNA with each fragment expressing different viral proteins. Influenza spreads seasonally, resulting in about three to five million yearly cases of severe illness and about 250,000 to 500,000 deaths annually or into the millions in pandemic years.
Specifications
| Influenza A H1N1 Virions | |
| Polyclonal | |
| HRP | |
| H1N1; H1N1 virions; Influenza H1N1 | |
| Influenza A, strain USSR (H1N1). | |
| 1 mL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1-2 mg/mL | |
| PBS with 10mg/mL BSA and 0.002% thimerosal; pH 7.2 | |
| Goat | |
| IgG fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.