missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INCENP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517201
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
Chromosomal passenger proteins are members of a group of proteins that move from centromeres to the spindle midzone during mitosis. INCENP (Inner Centromere Protein) is a chromosomal passenger protein with an essential role in mitosis and meiosis. It is a complex highly basic multidomain protein that is dynamically translocated from metaphase chromosomes to the spindle midzone during or just prior to anaphase. Some INCENP transfers to the equatorial cortex before formation of the cleavage furrow. Ultimately, it is discarded in the midbody at the completion of cytokinesis.
Specifications
| INCENP | |
| Polyclonal | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| Q9NQS7 | |
| Incenp | |
| Synthetic peptide corresponding to amino acids surrounding Pro240 of human INCENP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Incenp | |
| 2700067E22Rik; AU019509; binds and activates aurora-B and -C in vivo and in vitro; C130081E20; C77457; chromosomal passenger protein; Incenp; Inner centromere protein; inner centromere protein antigens 135/155kDa; inner centromere protein INCENP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3619 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction