missing translation for 'onlineSavingsMsg'
Learn More

ILK Mouse, Unlabeled, Clone: 3, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611802

 View more versions of this product

Product Code. 15826029

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Integrins are transmembrane receptors that mediate cell-cell or cell-matrix adhesion. All integrins are heterodimers composed of α and β subunits, which interact with extracellular matrix and cytoskeletal proteins. Signal transduction through intergin receptors may be regulated by integrin-linked kinase (ILK). ILK is a widely expressed Ser/Thr protein kinase that contains four ankyrin-like repeats in the N-terminal region, a phoshoinositide lipid-binding motif at amino acids 180-212, and an integrin binding site at amino acids 293-451. The ankyrin repeats interact with a LIM domain-only protein called PINCH that also binds the adaptor protein, Nck-2. This interaction implicates ILK in growth factor receptor pathways. In addition, the interaction of ILK with the cytoplasmic domains of integrin β1 and β3 subunits and localization of ILK to focal adhesion plaques suggests a role for ILK in the regulation of cell adhesion. Interestingly, overexpression of ILK in epithelial cells inhibits cell adhesion and stimulates cell cycle progression and tumorigenicity. Thus, ILK regulates signal transduction in many different cell signaling pathways.

Host Species: Mouse
Clone: 3
Isotype: IgG2
Species Reactivity [for Features Main]: Human
Immunogen: Human ILK aa. 326-452

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ILK
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human ILK aa. 326-452
50 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG1
Western Blot
3
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.