Learn More
interleukin enhancer binding factor 2, 45kDa, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ILF2.
Brand: Abnova H00003608-A01.50uL
Description
Nuclear factor of activated T-cells (NFAT) is a transcription factor required for T-cell expression of the interleukin 2 gene. NFAT binds to a sequence in the interleukin 2 gene enhancer known as the antigen receptor response element 2. In addition, NFAT can bind RNA and is an essential component for encapsidation and protein priming of hepatitis B viral polymerase. NFAT is a heterodimer of 45kDa and 90kDa proteins, the smaller of which is the product of this gene. The encoded protein binds strongly to the 90kDa protein and stimulates its ability to enhance gene expression. [provided by RefSeq
Sequence: PSEVLTMLTNETGFEISSSDATVKILITTVPPNLRKLDPELHLDIKVLQSALAAIRHARWFEENASQSTVKVLIRLLKDLRIRFPGFEPLTPWILDLLGHSpecifications
| interleukin enhancer binding factor 2, 45kDa | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ILF2. | |
| ILF2 | |
| MGC8391/NF45/PRO3063 | |
| Mouse | |
| 50 μL | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_004515 | |
| ILF2 | |
| ILF2 (NP_004506, 151 a.a. ∼ 250 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 3608 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.