missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL2RG Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA526461

Product Code. 13201479

  • £481.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IL2RG Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA), Flow

IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IL2RG
Polyclonal
Unconjugated
IL2RG
[g]c; Ab2-183; CD132; CD132 antigen; CIDX; common cytokine receptor gamma chain; cytokine receptor common subunit gamma; gamma(c); gammaC; gc; IL-2 receptor subunit gamma; IL2R gamma; IL-2R subunit gamma; IL2RG; IL-2RG; IMD4; interleukin 2 receptor subunit gamma; interleukin 2 receptor, gamma; interleukin 2 receptor, gamma (severe combined immunodeficiency); interleukin 2 receptor, gamma chain; Interleukin-2 receptor subunit gamma; p64; SCIDX; SCIDX1
Rabbit
Antigen affinity chromatography, Protein A
RUO
3561
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
0.35 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P31785
IL2RG
KLH conjugated synthetic peptide between 76-101 amino acids from the N-terminal region of human IL2RG.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.