missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL17D Polyclonal Antibody
IL17D Polyclonal Antibody
Brand: Invitrogen PA544831
This item is not returnable.
View return policy
Description
IL17D Polyclonal Antibody for Western Blot
Specifications
| IL17D | |
| Polyclonal | |
| Unconjugated | |
| IL17D | |
| AI462269; FLJ30846; IL17D; IL-17D; IL-22; IL-27; Il27a; IL-3R; Interleukin; interleukin 17D; interleukin 27A; interleukin-17D; Interleukin-27; UNQ3096/PRO21175 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 53342 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8TAD2 | |
| IL17D | |
| Synthetic peptide directed towards the N-terminal of human IL17D (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction