missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL1R1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529227
This item is not returnable.
View return policy
Description
Recommended positive controls: Molt-4, LNCaP. Predicted reactivity: Mouse (81%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The IL-1 system includes two agonists (IL-1 and IL-1), converting enzymes, antagonists, two receptors (IL-1 RI and IL-1 RII) and the IL-1 receptor accessory protein. Interleukin-1 signal is transduced through the type I receptor.
Specifications
| IL1R1 | |
| Polyclonal | |
| Unconjugated | |
| Il1r1 | |
| antigen CD121a; CD121 alpha; CD121 antigen-like family member A; CD121a; CD121b; D2S1473; il1 receptor; il-1 receptor; IL-1 receptor alpha chain; IL1R; il-1r; IL1R1; IL-1R1; IL-1R-1; Il1r-1; IL1RA; IL-1R-alpha; IL1RT1; IL-1RT1; IL-1RT-1; IL-iR; interleukin 1 beta receptor type 1 short transmembrane form; interleukin 1 beta receptor type 1 soluble form; interleukin 1 bete receptor type 1 long transmembrane form; interleukin 1 receptor alpha, type I; interleukin 1 receptor type 1; interleukin 1 receptor, type I; interleukin-1 receptor; interleukin-1 receptor alpha; Interleukin-1 receptor type 1; Interleukin-1 receptor type 1, membrane form; Interleukin-1 receptor type 1, soluble form; Interleukin-1 receptor type I; mIL-1R1; p80; sIL-1R1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3554 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P14778 | |
| Il1r1 | |
| Recombinant protein encompassing a sequence within the Intracellular domain of human IL1 Receptor 1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction