Learn More
IL-8 Mouse anti-Human, Unlabeled, Clone: G265-5, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554716
Description
The G265-8 antibody reacts with both the 72 and 77 amino acid forms of human interleukin-8 (IL-8). The immunogen used to produce the G265-8 hybridoma was recombinant human IL-8.
This antibody is routinely tested by ELISA as the capture. Other applications were tested at BD Biosciences Pharmingen during antibody development or reported in the literature.
Host Species: Rat
Clone: G265-5
Isotype: IgG2b κ
Species Reactivity: Human
Immunogen: Recombinant Human IL-8
ELISA, Western Blotting
Specifications
| IL-8 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human IL-8 | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG2b κ |
| ELISA | |
| G265-5 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.