Learn More
IL-7 Rat anti-Human, Biotin, Clone: BVD10-11C10, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554494
Description
The BVD10-11C10 antibody reacts with human interleukin-7 (IL-7). The immunogen used to generate the BVD10- 11C10 hybridoma was recombinant human IL-7. This is a non-neutralizing antibody.
Host Species: Rat
Clone: BVD10-11C10
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Recombinant Human IL-7
ELISA, Western Blotting
Specifications
| IL-7 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Recombinant Human IL-7 | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG2a |
| ELISA | |
| BVD10-11C10 | |
| Biotin | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.