Learn More
IL-6 Rat anti-Human, PE, Clone: MQ2-6A3, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 551473
Description
IL-6, a 26kDa protein, is a multifunctional cytokine secreted by both lymphoid and non-lymphoid cells. It regulates B and T cell function and hematopoiesis. The MQ2-6A3 antibody reacts with human interleukin-6 (IL-6). The immunogen used to generate the MQ2-6A3 hybridoma was recombinant human IL-6. This is a neutralizing antibody. Clone MQ2-6A3 cross-reacts with an intracellular component of peripheral blood LPS-stimulated permeabilized monocytes of rhesus and cynomolgus macaques. The reactivity pattern observed on non-human primates is similar, although weaker, than that seen on normal human monocytes.
Host Species: Rat
Clone: MQ2-6A3
Isotype: IgG2a κ
Species Reactivity: Non-human primate
Immunogen: Recombinant human IL-6
Flow Cytometry, Intracellular Staining
Specifications
| IL-6 | |
| Monoclonal | |
| PE | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| MQ2-6A3 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Recombinant human IL-6 | |
| 50 Tests | |
| Immunology | |
| Cynomolgus, Rhesus Monkey | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.