missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-3 Rat anti-Mouse, Unlabeled, Clone: MP2-8F8, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554381
This item is not returnable.
View return policy
Description
The MP2-8F8 antibody reacts with mouse interleukin-3 (IL-3). The immunogen used to generate the MP2-8F8 hybridoma was recombinant mouse IL-3. This is a neutralizing antibody.
Specifications
| IL-3 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant mouse IL-3 | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG1 |
| ELISA | |
| MP2-8F8 | |
| Unconjugated | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction