missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-2 Mouse anti-Human, Unlabeled, Clone: 5344.111, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555051
This item is not returnable.
View return policy
Description
The 5344.111 antibody reacts with human interleukin-2 (IL-2). The immunogen used to generate the 5344.111 hybridoma was affinity-purified, recombinant human IL-2 protein.
ELISA
Specifications
| IL-2 | |
| Monoclonal | |
| 0.5mg/mL | |
| IL2; Interleukin-2; T-cell growth factor; TCGF | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| ELISA | |
| 5344.111 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human IL-2 protein | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction